Mouse Recombinant G-CSF (from E. coli)
Supplier: Peprotech
AF-250-05-2UG AF-250-05-10UG AF-250-05-50UG AF-250-05-100UG AF-250-05-250UG AF-250-05-500UG AF-250-05-1MG
10771-740EA 289.23 CAD
10771-740 10771-742 10771-744 10771-746 10771-748 10771-750 10771-752
Mouse Recombinant G-CSF (from E. coli)
Proteins and Peptides
G-CSF is a hematopoietic growth factor that stimulates the development of committed progenitor cells to neutrophils and enhances the functional activities of the mature end-cell. It is produced in response to specific stimulation by a variety of cells, including macrophages, fibroblasts, endothelial cells and bone marrow stroma. G-CSF is being used clinically to facilitate hematopoietic recovery after bone marrow transplantation. Human and murine G-CSF is cross-species reactive. Recombinant Murine G-CSF is a 19.0 kDa protein consisting of 179 amino acid residues.Manufactured using all Animal-Free reagents.
  • Carrier free
  • Animal-free
MVPLVTVSAL PPSLPLPRSF LLKSLEQVRK IQASGSVLLE QLCATYKLCH PEELVLLGHS LGIPKASLSG CSSQALQQTQ CLSQLHSGLC LYQGLLQALS GISPALAPTL DLLQLDVANF ATTIWQQMEN LGVAPTVQPT QSAMPAFTSA FQRRAGGVLA ISYLQGFLET ARLALHHLA
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.