Human Recombinant IGF-II (from E. coli)
Supplier: Peprotech
AF-100-12-10UG AF-100-12-50UG AF-100-12-100UG AF-100-12-250UG AF-100-12-500UG AF-100-12-1MG
10771-694EA 304.45 CAD
10771-694 10771-696 10771-698 10771-700 10771-702 10771-704
Human Recombinant IGF-II (from E. coli)
Proteins and Peptides
The IGFs are mitogenic, polypeptide growth factors that stimulate the proliferation and survival of various cell types, including muscle, bone, and cartilage tissue in vitro . IGFs are predominantly produced by the liver, although a variety of tissues produce the IGFs at distinctive times. The IGFs belong to the Insulin gene family, which also contains insulin and relaxin. The IGFs are similar to insulin by structure and function, but have a much higher growth-promoting activity than insulin. IGF-II expression is influenced by placenta lactogen, while IGF-I expression is regulated by growth hormone. Both IGF-I and IGF-II signal through the tyrosine kinase type I receptor (IGF-IR), but IGF-II can also signal through the IGF-II/Mannose-6-phosphate receptor. Mature IGFs are generated by proteolytic processing of inactive precursor proteins, which contain N-terminal and C-terminal propeptide regions. Recombinant Human IGF-I and IGF-II are globular proteins containing 70 and 67 amino acids, respectively, and 3 intra-molecular disulfide bonds. The calculated molecular weight of Recombinant Human IGF-II is 7.5 kDa.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.