Yeast Recombinant Kex-2 (from Hi-5 Insect Cells)
Supplier: Peprotech
450-45-50UG 450-45-250UG
10771-124EA 226.59 CAD
10771-124 10771-126
Yeast Recombinant Kex-2 (from Hi-5 Insect Cells)
Proteins and Peptides
Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to, specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, as they are frequently used in the analysis and production of proteins. Kex-2 cleaves at the carboxyl end of the recognition sequences Arg-Arg/X and Lys-Arg/X. Recombinant Yeast Kex-2 is a 60.4 kDa protease consisting of 558 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.