Human Recombinant Galectin-1 (from E. coli)
Supplier: Peprotech
450-39-10UG 450-39-50UG 450-39-100UG 450-39-250UG 450-39-500UG 450-39-1MG
10771-086EA 209.84 CAD
10771-086 10771-088 10771-090 10771-092 10771-094 10771-096
Human Recombinant Galectin-1 (from E. coli)
Proteins and Peptides
Lectins, of either plant or animal origin, are carbohydrate-binding proteins that interact with glycoprotein and glycolipids on the surface of animal cells. The Galectins are lectins that recognize and interact with beta-galactoside moieties. Galectin-1 is an animal lectin that has been shown to interact with CD3, CD4, and CD45. It induces apoptosis of activated T-cells and T-leukemia cell lines, and inhibits the protein phosphatase activity of CD45. Recombinant Human Galectin-1 is a 14.5 kDa protein containing 134 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.