Mouse Recombinant LIGHT (from E. coli)
Supplier: Peprotech
315-12-5UG 315-12-20UG 315-12-100UG 315-12-250UG 315-12-500UG 315-12-1MG
10770-926EA 209.84 CAD
10770-926 10770-928 10770-930 10770-932 10770-934 10770-936
Mouse Recombinant LIGHT (from E. coli)
Proteins and Peptides
LIGHT belongs to the TNF family of ligands, and can signal through the herpes virus entry mediator type A receptor (HVEM, TNFRSF14), LTβR, or bind to a decoy receptor, DcR3. It is expressed in splenocytes, activated PBL, CD8+ tumor infiltrating lymphocytes, granulocytes, and monocytes. LIGHT has the ability to active NF-κB, to co-stimulate the activation of lymphocytes and to induce apoptosis in certain human tumor cells. Recombinant Murine LIGHT is a 20.1 kDa protein containing 183 amino acid residues, comprising the TNF-homologous region of the LIGHT extracellular domain.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.