Mouse Recombinant EGF (from E. coli)
Supplier: Peprotech
315-09-100UG 315-09-500UG 315-09-1MG
10770-906EA 230.96 CAD
10770-906 10770-908 10770-910
Mouse Recombinant EGF (from E. coli)
Proteins and Peptides
EGF is a potent growth factor that stimulates the proliferation of various epidermal and epithelial cells. Additionally, EGF has been shown to inhibit gastric secretion, and to be involved in wound healing. EGF signals through a receptor known as c-erbB, which is a class I tyrosine kinase receptor. This receptor also binds with TGF-α and VGF (vaccinia virus growth factor). Recombinant Murine EGF is a 6.0 kDa globular protein containing 53 amino acid residues, including 3 intramolecular disulfide bonds.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.