Human Recombinant sCD100 (from CHO Cells)
Supplier: Peprotech
310-29-5UG 310-29-20UG 310-29-100UG
10770-898EA 209.84 CAD
10770-898 10770-900 10770-902
Human Recombinant sCD100 (from CHO Cells)
Proteins and Peptides
The Semphorins are a large family of phylogenetically conserved proteins that play a pivotal role in maintaining homeostasis in the immune system. Twenty members of this family have been identified and categorized into eight subclasses based on sequence similarity and distinctive structural features. CD100, also known as Sema4D, is a 150 kDa transmembrane class IV semaphorin. Studies have shown that CD100 can induce monocyte migration, T-cell activation, and B-cell survival, as well as T/B cell and T/DC “cooperation”. The CD100 precursor contains 862 amino acids, including a 21 a.a. signal sequence, a 713 a.a. extracellular domain, a 21 a.a. transmembrane sequence, and a 107 a.a. cytoplasmic region. The extracellular sequence contains several structural features, including a 479 a.a. “sema” domain, a 79 a.a. Ig-like sequence, and a 52 a.a. “Plexin-type repeat”. Recombinant Human sCD100 is a 78.9 kDa protein comprising the extracellular domain of CD100 (711 amino acids). SDS-PAGE analysis run under non-reducing conditions shows a mixture of disulfide linked dimer and monomer.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.