Human Recombinant HVEM-Fc (from (BTI-Tn-5B1-4) Hi-5 Insect cells)
Supplier: Peprotech
310-27-20UG 310-27-100UG 310-27-500UG
10770-876EA 221.22 CAD
10770-876 10770-878 10770-882 10770-884
Human Recombinant HVEM-Fc (from (BTI-Tn-5B1-4) Hi-5 Insect cells)
Proteins and Peptides
HVEM belongs to the TNF Receptor superfamily of transmembrane proteins, and plays a role in the activation of T-cells and other lymphocytes. It is expressed in various cells and tissues, including spleen, thymus, lung, macrophages, and T-cells. HVEM activation induces a signaling cascade that results in the induction of transcription factors NF-κB and AP-1. LIGHT (TNFSF14) and TNF-β (TNFSF1) function as the ligands for HVEM, which can also bind specifically to herpes simplex virus glycoprotein D. Soluble HVEM can act as a “receptor decoy” resulting in inhibition of the activity of the HVEM ligands, LIGHT and TNF-β. Recombinant Human HVEM-Fc is a 376 amino acid fusion protein that contains an N-terminal domain corresponding to the extracellular region of HVEM, and a C-terminal domain corresponding to residues 102 to 330 of human IgG1. The calculated molecular weight of Recombinant Human HVEM-Fc is 41.4 kDa.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.