Human Recombinant sCD23 (from E. coli)
Supplier: Peprotech
310-26-5UG 310-26-20UG 310-26-100UG 310-26-250UG 310-26-500UG 310-26-1MG
10770-864EA 209.84 CAD
10770-864 10770-866 10770-868 10770-870 10770-872 10770-874
Human Recombinant sCD23 (from E. coli)
Proteins and Peptides
CD23, the low affinity receptor for IgE, belongs to the C-type lectin structural family and plays a role in the regulation of IgE synthesis and IgE mediated activities. It is found both as a transmembrane receptor protein and in a soluble form, which is generated by proteolytic cleavage of membrane-bound CD23. The predominant soluble form of CD23 (sCD23) consists of 172 amino acids corresponding to the extracellular domain of the full length precursor. sCD23, in addition to binding IgE, also exerts a number of IgE-independent activities, such as promoting the activation and differentiation of B-cells and stimulating the release of pro-inflammatory cytokines from monocytes. Recombinant Human sCD23 is a 19.2 kDa non-glycosylated protein containing 172 amino-acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.