Human Recombinant Eotaxin-3 (from E. coli)
Supplier: Peprotech
300-48-500UG 300-48-1MG
10770-720EA 4381.56 CAD
10770-720 10770-722
Human Recombinant Eotaxin-3 (from E. coli)
Proteins and Peptides
Eotaxin-3 is a CC chemokine that signals through the CCR3 receptor. It is produced by endothelial cells stimulated with IL-4 or IL-13. Eotaxin-3 selectively targets cells expressing CCR3, including eosinophils, basophils, T cells and monocytes. Eotaxin-3 has similar activity to Eotaxin and Eotaxin-2, but the three Eotaxins share only a low degree of sequence homology. Recombinant Human Eotaxin-3 (CCL26) is an 8.4 kDa protein containing 71 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.