Human Recombinant PlGF-1 (from E. coli)
Supplier: Peprotech
100-06-5UG 100-06-25UG 100-06-100UG 100-06-250UG 100-06-500UG 100-06-1MG
10770-666EA 209.84 CAD
10770-666 10770-668 10770-670 10770-672 10770-674 10770-676
Human Recombinant PlGF-1 (from E. coli)
Proteins and Peptides
PlGF-1 is an angiogenic factor that belongs to the cysteine-knot superfamily of growth factors. PlGF-1 is expressed in placental tissues, the colon, and mammary carcinomas. It signals through the VEGFR-1/FLT1 receptor, and stimulates endothelial cell proliferation and migration. Recombinant Human PlGF-1 is a 29.7 kDa disulfide-linked homodimeric protein of two 132 amino acid polypeptide chains.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.