Anti-IL18BP Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Avantor
10392-020EA 1281.54 CAD
10392-020
Anti-IL18BP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Isoform A binds to IL-18 and inhibits its activity. Functions as an inhibitor of the early TH1 cytokine response.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL18BP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR