VWR® TraceClean® Vials, Clear
Supplier: VWR International
89093-840 89096-790 89126-532 89126-534 89126-538 89126-540 89126-542
89093-840CS 866.99 CAD
89093-840 89096-790 89126-532 89126-534 89126-538 89126-540 89126-542
VWR® TraceClean® Vials, Clear
Vials
These TraceClean® glass jars are used for the collection of and testing of aqueous samples for Volatile Organic Analysis and Total Petroleum Hydrocarbons (TPH).
  • Clear glass fabrication
  • Suited for testing aqueous samples
  • Used as a containment vessel
Collection and testing of aqueous samples for Total Organic Carbon (TOC)
Quality Assured® with VWR® logo sample ID label products, a lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement. This certificate of analysis is included in each product case.
Secondary uses - Containment of extracts, etc. Please refer to the applicable certificate of analysis for certified analytes and corresponding reporting limits.
Certifications: QAL and PC (89096-790)
Ordering information: Container measurements and capacity 'contain' volumes may vary.