VWR® TraceClean® Straight Side Glass Jars, Clear
Supplier: VWR International
89126-568 89126-570 89126-578 89126-580 89126-582 89126-584 89126-592 89126-596 89094-235
89126-568CS 307.06 CAD
89126-568 89126-570 89126-578 89126-580 89126-582 89126-584 89126-592 89126-596 89094-235
VWR® TraceClean® Straight Side Glass Jars, Clear
Jars General Use Jars
These TraceClean® glass jars are used primarily for the collection of soil, sediments, and slurry media for testing Semi-volatiles, Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics.
  • Clear glass fabrication
  • Straight-sided design
  • Used as a containment vessel
  • Suited to the collection of inorganic samples
Secondary uses include containment, bench vessels, and archiving
The 950 mL clear jar 89094-235 contains 5 mL (1:1) of the chemical preservative H₂SO₄.
Quality Assured® with VWR® Logo Sample ID Label products, a lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement. This certificate of analysis is included in each product case.
Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Certifications: QAL.
Ordering information: Container measurements and capacity "contain" volumes may vary.