VACUUBRAND VACUU·VIEW and VACUU·VIEW Extended Vacuum Gauges, BrandTech
Supplier: Brandtech
20683220 20683210
10024-956EA 3536.43 CAD
10024-956 10024-958
VACUUBRAND VACUU·VIEW and VACUU·VIEW Extended Vacuum Gauges, BrandTech
Pumps Vacuum Gauges
Vacuum gauges with corrosion resistant transducers designed especially for the rigors of laboratory use
  • Mercury-free
  • Digital display for accurate readings
  • Capacitive and Pirani transducers for continuous reading from atmosphere to 10-3 mbar/Torr/hPa
  • Corrosion resistant transducers
The VACUU·VIEW gauge features a capacitive transducer made of alumina ceramic, and is suitable for absolute pressure readings in the rough vacuum range (atmosphere to 0.1mbar/hPa or 0.075Torr). Suitable for rotary evaporators, or any application performed by diaphragm, roots or scroll pumps as well as house vacuum.
The VACUU·VIEW extended features a dual technology gauge head, a capacitive transducer for absolute pressure readings in the rough vacuum range, paired with a Pirani transducer for measurements in the fine vacuum range. Both transducers have alumina ceramic wetted surfaces for corrosion and shock resistance. Suitable for freeze drying, short path distillation, and other applications where rotary vane vacuum pumps are used.
Both gauges feature KF16 small flange vacuum connections, with screw-in hose barbs to allow fitting of 6-10mm ID vacuum hose. VACUU·BUS connections, allow interfacing with VACUUBRAND DCP3000 and CVC3000. Universal power supplies included.
Ordering information: Supplied complete with operating manual and 100-240V power supply with US, CEE, UK, CH and CN plugs.