Merit™ Operating Scissors, Physician Grade, Sklar
Supplier: Sklar
97-295 97-299 97-263 97-266 97-269 97-281 97-284 97-272 97-270 97-275 97-278 97-292
10184-404EA 22.31 CAD
10184-404 10184-406 10184-600 10184-602 10184-604 10184-606 10184-608 10193-642 10193-682 10193-704 10193-872 10194-480
Merit™ Operating Scissors, Physician Grade, Sklar
Scissors
Merit™ operating scissors, also known as surgical scissors are a versatile surgical instrument
Merit™ instruments are office-grade line of products used for routine and in-office minor procedures.
These high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit™ instruments deliver a solution for physicians and practices that require quality, durable instruments.
They are used for general, multipurpose cutting and dissecting. Sharp operating scissors cut tougher tissue and are frequently used for dissecting, whereas blunt operating scissors areMore Product Information