Certis® Ready Engineered Shippers, Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
CDUS3XE CRUS2LP CCUS3XEEP CCUS4XEEP CCUS3LP CRUG3XP CRUG3SP CRUG5LV C6R5U C6C5U C6F5U C26R5U C26C5U C26F5U C14F5U C14R5U C14C5U C6R3U C6C3U C6F3U C14R3U C14C3U C14F3U C26R3U C26C3U C26F3U C56R5U C56C5U C56R3U C56C3U C56F3U C13C2U C13F2U C13R2U C27C2U C27F2U C27R2U C56C2U C56F2U C56R2U C7C2U C7F2U C7R2U C3R3U CRUS2XE
10218-606PL 2040.58 CAD
10218-606 10218-774 10770-540 10770-542 10770-546 89213-069 89213-071 89213-075 10841-240 10841-242 10841-244 10841-246 10841-248 10841-250 75788-502 75788-516 75788-518 75817-428 75817-430 75817-432 75817-434 75817-436 75817-438 75817-440 75817-442 75817-444 75839-352 75839-354 75839-356 75839-368 75839-370 75839-372 75959-622 75959-624 75959-626 75959-628 75959-630 75959-632 75959-634 75959-636 75959-638 75959-640 75959-642 75959-644 76204-640 89213-101
Certis® Ready Engineered Shippers, Sonoco ThermoSafe
Mailers and Shippers
Robust, universal insulated shipping solutions optimized to provide the lowest total cost of ownership available.
  • One-way universal solutions
  • Operational efficiency, redefined
  • Material neutral approach
  • PureTemp® vegetable-based phase change material
Our Certis line of premium pre-qualifed insulated shippers are ideal for one-way shipments of pharmaceutical products across summer or winter seasons for refrigerated, CRT and frozen shipping solutions from 48 hours to 120 hours of duration.
Material neutral approach utilizes leading edge technologies to provide the lowest total cost of ownership. Designed solutions reduce the amount of weight, packout complexity, and overall size, resulting in reduced shipping costs. Designs feature PureTemp® Vegetable-Based Phase Change Material, which is a natural phase change material (PCM) that provides strict temperature control and greater efficiency, resulting in packages that are smaller and lighter.
The Certis Platform of shippers are designed to offer operational efficiency and ease of personnel training using ZERO Bench-Time® technology and same pack-out configuration across durations. Certis Platform utilizes the same pack-out for refrigerated, CRT and frozen solutions from 48 hours to 120 hours of duration. These shippers optimize inventory and provide lowest total cost of ownership utilizing only 3 types of bricks and 8 shippers to create 24 shipping solutions across temperature ranges and duration.
Designed to a variety of different ambient temperature profiles representative of moderate to extreme external distribution environments.
Certifications: Pre-Qualified with the choice of two different ambient profiles, ISCgold® and ISCsilver®.
Ordering information: Models CCUS3XEEP, CCUS4XEEP, and CCUS3LP are shippers only. Contact your Avantor representative for refrigerant components, which are sold separately.