Metzenbaum Dissecting Scissors, Floor Grade, Sklar
Supplier: Sklar
21-335 21-336 21-339 21-340
89023-156PK 148.78 CAD
89023-156 89023-158 89023-162 89023-164
Metzenbaum Dissecting Scissors, Floor Grade, Sklar
Scissors
Metzenbaum Dissecting Scissors are used for dissecting and cutting tissue.
They are not recommended for cutting sutures, drains, or heavy tissue.