Sklarlite XD™ Iris Scissors, OR Grade, Sklar
Supplier: Sklar
23-1155 23-1154 23-1152 23-1153
10193-910EA 379.59 CAD
10193-910 10194-018 10194-344 10194-346
Sklarlite XD™ Iris Scissors, OR Grade, Sklar
Scissors
Sklarlite XD™ Iris Scissors are light and delicate instruments, which allow a high degree of precision with the least amount of fatigue, in emergency rooms, for ophthalmic, OB/GYN and dermatologic procedures.
They have become a multipurpose instrument used in various surgical environments. These fine, sharp, finger ring scissors are designed for detailed dissection of fine tissue and fine suture removal.
Order Now
SPECIFICATIONS
Environmentally Preferable