Econo™ Spencer Stitch Scissors, Sklar
Supplier: Sklar
21-352 96-2596 96-2598 96-2663 96-2672
89023-170PK 131.35 CAD
89023-170 89023-934 89023-938 89023-984 75995-416
Econo™ Spencer Stitch Scissors, Sklar
Scissors
Econo™ Spencer Stitch Scissors are primarily used for suture removal.
  • Small hook-shaped tip on one blade
  • Delicate tip for removing smaller sutures
These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They typically have smaller, more delicate tips that are used for removing smaller sutures. These scissors should not be used for cutting tissue.