Wagner Scissors, OR Grade, Sklar
Supplier: Sklar
47-1047 47-1048 47-1049 47-1058 47-1059 47-1068 47-1075 23-1254 47-1069 47-1070 47-1071
89024-914EA 241.46 CAD
89024-914 89024-916 89024-918 89024-920 89024-922 89024-926 89024-934 10195-278 10196-798 10196-824 10196-826
Wagner Scissors, OR Grade, Sklar
Scissors
Wagner Scissors are finger ring surgical scissors used in procedures requiring delicate or precise cutting, particularly plastic surgery.
Curvature, blade, and tip style can be tailored to meet the needs of specific procedures.