Iris Scissors, OR Grade, Sklar
Supplier: Sklar
47-1135 47-1140 47-1144 47-1145 47-1235 47-1240 47-1244 47-1245 47-1246 47-1248 47-1440 47-1441 22-1135 22-1235 47-1143 47-1243 47-1442 64-2142 64-2042
82030-230EA 176.89 CAD
82030-230 82030-232 82030-234 82030-236 82030-238 82030-240 82030-242 82030-244 82030-246 89024-952 89024-962 89024-964 89025-212 89025-228 10194-356 10194-358 10194-360 10194-362 10194-364 10197-872
Iris Scissors, OR Grade, Sklar
Scissors
Iris Scissors are designed for ophthalmic procedures, these fine and sharp scissors are suitable for detailed dissection of fine tissue and fine suture removal.
  • Multipurpose instrument used in various surgical environments
In addition to ophthalmic procedures, Iris scissors are used in emergency rooms and for obstetrics and gynecology as well as dermatologic procedures.
The Ready OR Grade Iris Scissors are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.