ADC® Satin Laryngoscope Set
Supplier: AMERICAN DIAGNOSTIC CORP CP
ADC®
4089
470119-674CS 4756.05 CAD
470119-674
ADC® Satin Laryngoscope Set
Educational Materials Biology Educational Materials Human Biology Learning Activities
Full assortment of blades and handles in a fitted case.
  • Easy to store fitted case
  • Compatible with major brands of fiber-optic handles or blades
This set of satin handles and miller blades is a great way to keep your set organized. All of the components easily store in a rigid case with die cut foam inserts. Each of the two handles (C size and AA) are chrome plated brass with a knurled finish giving a good grip even with wet gloves. The blades are made of stainless steel for easy cleaning and service.
Blades and Handles are compatible with other brands conforming with ASTM F965, or ISO 7376.
Accessories information: Does not include batteries.
Delivery information: Includes 2 handles, 5 blades and 1 year warranty.