Sims Scissors, OR Grade, Sklar
Supplier: Sklar
90-1080 90-1180 90-1280 90-1380 90-1480 90-1580
10194-242EA 301.04 CAD
10194-242 10197-154 10197-156 10197-162 10197-164 10197-166
Sims Scissors, OR Grade, Sklar
Scissors
Sims scissors are finger ring instruments, that feature long blades for deep tissue dissection.
Ordering information: Available in curved tip style.