4-(Maleimidomethyl)benzonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

H58015-03 H58015-06
AAH58015-03EA 246.48 CAD
AAH58015-03 AAH58015-06
4-(Maleimidomethyl)benzonitrile 97%
4-(Maleimidomethyl)benzonitrile
Formula: C₁₂H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD16665080
CAS Number: 1249094-81-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR