GelJacket™ Stackable CO₂ Incubators, 7404 Series, Caron Products

Supplier: Caron Products
7404-10-2 7404-10-3 7404-10-1
10158-770EA 41978.34 CAD
10158-770 10158-772 10158-852
GelJacket™ Stackable CO₂ Incubators, 7404 Series, Caron Products
Incubators CO₂ Incubators
Caron’s GelJacket is a high-volume, controlled humidity CO2 incubator that replaces conventional water jacketed units as the benchmark for critical culture protection.

GelJacket offers precise gVapor™ controlled humidification, as well as a rapid 90° decon cycle for fast chamber cleaning. These standard features save energy and user time, as opposed to contamination-prone water pan-based systems and high temperature or complicated competing decontamination processes.

Refrigerated models with extended temperature ranges are available for applications that generate heat (such as shakers).More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable Sustainable Packaging,Energy Efficient
CO₂ range 0 - 20%, +/- 0.1%
Temperature uniformity +/- 0.3°C
Temperature sensor RTD
Temperature control +/-0.1°C
Int. dimensions 58.4W x 65.6D x 75.7H cm (23 x 25.8 x 29.8")
Humidity range Ambient to 95%
Humidity control gVapor active control (Humidification System)
Exterior dimensions 112.3W x 82.8D x 92.7H cm (44.5 x 31.7 x 37")
CO₂ sensor IR

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR