Bouin's Solution, Azer Scientific
Supplier: Azer Scientific
B-F-G CUNB-5G CUNB-2.5G
89217-520EA 660.77 CAD
89217-520 89217-516 89217-518
Bouin's Solution, Azer Scientific
Stains Microscopy and staining reagents
Bouin's solution is a saturated picric acid-formalin fixative that is useful for preserving delicate tissues and gastrointestinal tract specimens.
Packaging: Dispending spigots are included with cube containers.