Prefilled Bouin's Solution, Azer Scientific
Supplier: Azer Scientific
PFB-020 PFB-040 PFB-060
89217-510CS 459.6 CAD
89217-510 89217-513 89217-515
Prefilled Bouin's Solution, Azer Scientific
Cell Culture Supplements and Additives
Bouin's fixative is a saturated picric acid-formalin solution that is useful for preserving and fixing delicate tissues and gastrointestinal tract specimens.
  • Available in 20, 40 and 60 ml containers each half-filled
  • 95 kPa compliant specimen containers