Patient Control Slide, Azer Scientific
Supplier: Azer Scientific
ESCS-100 ESCS-101
89176-820CS 1972.88 CAD
89176-820 89176-822
Patient Control Slide, Azer Scientific
Slides Microscopy Control Slides
The patient control slide is suitable for staining protocols that require a control.
  • Spaces for Both Patient Sample and Control Material
  • Chemical Adhesive for Better Tissue Adhesion
  • Red Line on Reverse Side of Slide for Unrestricted Movement of Reagents
Each slide features space for both patient sample and control material
Slide is positively charged for better tissue adhesions. Available without clipped corners or with clipped corners for use in automated systems.
The slide is positively charged for better tissue adhesions. Available without clipped corners or with clipped corners for use in automated systems.
A red line allows for the two samples to be separated. This red line is printed on reverse side of slide to allow for unrestricted movement of reagents.