Corning® LSE™ 6 Liter Digital Water Bath, Corning
Supplier: Corning
6783 6784
10020-984CS 1931.28 CAD
10020-984 10020-986
Corning® LSE™ 6 Liter Digital Water Bath, Corning
Baths Water Baths, Unstirred
The Corning LSE Digital Water Bath is a personal-sized bath constructed from corrosion resistant stainless steel
  • Compact design
  • Stainless steel chamber and cover
  • Powerful 500W heater programmed by LED digital controller
  • Precise temperature control by hydraulic thermostat
The seamless chamber can accommodate up to 6 liters of water or can serve as a dry bath when using thermal beads. Temperature can be set from between ambient +5°C and 99°C and easily monitored on the digital display. A stainless steel gable cover is also included, which helps retainMore Product Information