SMZ-171 Stereo Microscope, Motic
Supplier: Motic
1100200600758 1100200600754 1100200600762 1100200600767
CA10769-598EA 2354.6 CAD
CA10769-598 CA10769-600 CA10769-602 CA10769-604
SMZ-171 Stereo Microscope, Motic
Microscopes
Flexible stereo microscopes for advanced research applications in biomedicine and material science. The 6.7:1 Greenough zoom delivers a standard magnification of 7.5x - 50x with 110 mm working distance in a FOV23mm.
  • Eyepieces feature Widefield N-WF10X/23 mm with diopter adjustment
Delivery information: Supplied with the following: Stands, Stage Accessories, Micrometer Reticles, Additional Illumination, Polarizing Equipment, Darkfield Equipment, Barlow Lenses. Microscope is customizable with various accessories. For all available options, please contact your local Avantor sales representative.