Motic BA310 Elite Upright Compound Microscope
Supplier: Motic
89168-482 1101001100401 1100100402631 1100100402641
89168-482EA 133.6 CAD
89168-482 CA89168-482 CA10769-642 CA10769-646 CA10769-650
Motic BA310 Elite Upright Compound Microscope
Microscopes
Motic’s BA310 Elite Series is built for the performance demands of colleges, universities, and medical school markets, setting a new price-performance standard for optical quality
The BA310 Elite model introduces superior improvements in both optical and mechanical performance. The rackless stage concept without a prominent gear rack permits even more convenient x/y movement and effortless positioning of the stage without gear rack interference. Ergonomic drop-down coaxial controls enable convenient 'eyes down' operation for uninterrupted sample viewing. The included CCIS EC PlanMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Eyepieces WF 10×/20 mm WF 10×/20 mm
Objectives CCIS EC plan achromatic EC PL 4×, 10×, 40× S, 100× S-oil CCIS EC plan achromatic EC PL 4×, 10×, 40× S, 100× S-oil
Magnification 40x to 100x 40 - 1000×
Condenser Abbe with Iris Diaphragm and Slot (NA = 0.9/1.25) Abbe with Iris Diaphragm and Slot (NA = 0.9/1.25)
Illumination Halogen or LED (optional) Halogen or LED (optional)
Electrical 100–240V 100–240V
Optical design Infinity Corrected Infinity Corrected
Accessories included Micrometer Reticles, Phase Contrast Accessories, Polarizing Accessories, Darkfield Accessories