Accessories for KIMBLE® MICROFLEX® Williamson Microscale Kits, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
746001-0001 748001-0003 748001-1000 748003-0000 748003-1000 748004-1000 748005-0000 748005-1000 748005-1001 748005-1651 748006-1100 748007-1060 748008-1005 748009-0005 748010-1025 748012-1000 748013-1000 748014-0000 748014-1000 748017-0020 748020-1000 748025-1034 764100-1001 748002-0000 748004-0000 748006-0100 748008-0005 748010-0025 748012-0000 748013-0000 748030-0049
KT746001-0001CS 88.05 CAD
KT746001-0001 KT748001-0003 KT748001-1000 KT748003-0000 KT748003-1000 KT748004-1000 KT748005-0000 KT748005-1000 KT748005-1001 KT748005-1651 KT748006-1100 KT748007-1060 KT748008-1005 KT748009-0005 KT748009-1005 KT748010-1025 KT748012-1000 KT748013-1000 KT748014-0000 KT748014-1000 KT748017-0020 KT748020-1000 KT748025-1034 KT748033-0008 KT764100-1001 KT748001-0000 KT748002-0000 KT748004-0000 KT748006-0100 KT748008-0005 KT748010-0025 KT748012-0000 KT748013-0000 10024-800
Accessories for KIMBLE® MICROFLEX® Williamson Microscale Kits, DWK Life Sciences
Microchemistry Kits
Test fabric is useful for testing dyes on a variety of fabrics
Three-foot length of fabric four inches wide features 8 strips of different fibers woven side by side. Filament acetate, bleached cotton, spun polyamide type 66, polyester disperse dyeable, spun polyacrylic, spun silk, spun viscose, and worsted wool.
Accessory for Microscale Williamson kit.
Order Now
SPECIFICATIONS
Environmentally Preferable