Limited Volume Vial Insert, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
60850-528 60850-629 60855-531 60855-631
10024-788CS 175.66 CAD
10024-788 10024-790 10024-792 10024-794
Limited Volume Vial Insert, Kimble Chase
Vials
Inserts are ideal for limited and micro volume applications.
  • Manufactured from clear borosilicate glass
Options are available for both standard and wide opening 12×32 autosampler type vials.
60850 series has a conical point design with polymer springs attached and residual volume of 1 µl. 60855 series has a flat bottom design and residual volume of 25 µl.