Motic SMZ-161 Stereo Microscope
Supplier: Motic
89168-448 1101000300641 1101000300452 1101001401861 1101001401841 1101001400141
89168-444EA 85.32 CAD
89168-444 89168-446 89168-448 CA19000-196 CA19000-666 CA89168-444 CA89168-446 CA89168-448
Motic SMZ-161 Stereo Microscope
Microscopes
Employing the Greenough stereoscopic optical system, the Motic SMZ-161 series of stereo microscopes provides large-scale zoom ratio and the clearest image, with high distortion-free resolution and depth of field for sharp, 3-dimensional images
The Greenough optical system delivers clear and distortion-free images with an improved zoom range of 6:1. Parfocality is ensured while running through the complete magnification range; a click-stop mechanism, implemented in the continuous zoom, enables precise reproduction of magnifications. The extended 110mm working distance in itsMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Eyepieces
Illumination
Electrical
Optical design