Ovation® M, Ergonomic Mechanical Pipettors, VistaLab
Supplier: VistaLab
1070-0002 1070-0010 1070-0020 1070-0100 1070-0200 1070-1000 1070-0998 1070-0999
10018-836EA 565.25 CAD
10018-836 10018-838 10018-840 10018-842 10018-844 10018-846 10018-848 10018-850
Ovation® M, Ergonomic Mechanical Pipettors, VistaLab
Pipettes Micropipettes
Ovation® pipettors have a unique and patented ergonomic design, the only type that will keep user's hand and wrist in the neutral position recommended by ergonomic experts.
The angled shape allows users to pipette with low arm and elbow height, decreasing tension and associated pain in the neck, shoulder, back, and elbow. The Ovation® M Mechanical model combines conventional analog volume setting with VistaLab Technologies superior accuracy and precision. The contoured shape and adjustable hook assures a comfortable, easy grip and custom fit hand hold. The spring loaded tip acquisition means a gentle push is all that is required to attach tips, and an audible 'click' confirms secure attachment. Tips 'flick' off easily with a low force push of the eject button.
Special filters in the nozzle prevent liquid from being inadvertently aspirated into the pipettor, protecting it from contamination. Ovation pipettors are the only pipettors that stand on their own, which means tips never touch the bench, even if the pipettor is put down in the middle of a procedure. These devices are color-coded to make identification quick and easy for volume range selection. A convenient microtube cap-opener is built in to the base. Ovation® M pipettors are fully autoclavable.
Ordering information: Ovation® M pipettors are supplied with a two-year warranty.