MINUM-WARE®, Neoprene Filter Adapters, Chemglass
Supplier: Chemglass
MW-121-01 MW-121-02
80064-014EA 13.79 CAD
80064-014 80064-016
MINUM-WARE®, Neoprene Filter Adapters, Chemglass
Supports Support Sleeves
Microscale neoprene filter adapters for use with MW-122 filtering flasks.
Adapters may be nested to permit use of funnels with a stem diameter as small as 6 mm O.Ø.