96-Well Preparation Rack with Cover, Bio Plas
Supplier: Bio Plas
0034F 0035F 0032F 0031F 0030F 0033F
21150-222CS 909.43 CAD
21150-222 21150-220 21150-216 21150-226 21150-224 21150-218
96-Well Preparation Rack with Cover, Bio Plas
Racks Tube Racks
High quality
  • Designed for preparation and storage
  • Racks for use with a variety of microcentrifuge tubes
Manufactured of polypropylene, these 96-well (8x12 format) racks are suitable for autoclaving. These one sided racks are suitable for 0.2 ml PCR tubes with strip caps. The snap-on cover stays in place during transportation and storage. Racks are able to be securely stacked.
Made in the USA.