Malonyl coenzyme A lithium salt ≥95%, powder
Supplier: CHEM-IMPEX INTERNATIONAL, INC
01888-25MG
01888-5MG
CH01888-25MGEA
822.67
CAD
CH01888-25MG
CH01888-5MG
Malonyl coenzyme A lithium salt ≥95%, powder
Malonyl coenzyme A lithium salt
Formula:
C₂₄H₃₇N₇O₁₉P₃SLi MW: 859.6 g/mol |
MDL Number:
MFCD00057769 CAS Number: 108347-84-8 |
Specification Test Results
Purity Limit | ≥ 90% (HPLC) |
Molecular Formula | C24H37N7O19P3SLi |
Molecular Weight | 859.6 |
CAS | 108347-84-8 |
MDL | MFCD00057769 |
Apperance | White powder |
Storage Temperature | Store at ≤ -20 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...