Septum Stoppers, Sleeve Type, Chemglass
Supplier: Chemglass
CG-3022-91 CG-3022-97 CG-3022-93 CG-3022-09 CG-3022-99 CG-3022-96 CG-3022-95 CG-3022-98
89029-092EA 48.37 CAD
89029-092 33501-388 33501-630 80062-432 80062-434 10170-486 10170-488 10170-490 10170-492
Septum Stoppers, Sleeve Type, Chemglass
Stoppers Rubber Stoppers
Sleeve type stoppers with hollow plugs and sleeve extensions.
  • Available in red or white rubber
The sleeve extensions fold down over the neck of a vessel and tubing. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.