Whizard® Liner II Gloves, Wells Lamont
Supplier: Wells Lamont
333132 333134 333136 333278
80090-871EA 69.38 CAD
80090-871 80090-873 80090-875 80090-879
Whizard® Liner II Gloves, Wells Lamont
Gloves
The Whizard Liner II gloves provide comfort and softness and made of Spectra®, fiberglass and stainless steel.
  • Wear alone or as a liner
  • Ambidextrous
  • Gray to better conceal stains
  • Medium duty
Suitable for metal stamping, parts handling, glass handling, sheet rolling, rubber trimming and plastic trimming applications.