Explorer® Semi-Micro Balances, OHAUS®
Supplier: Ohaus
30139512 30139513 30139514 30139510 30139511
10013-890EA 16621.52 CAD
10013-890 10013-892 10013-894 10013-898 10013-900
Explorer® Semi-Micro Balances, OHAUS®
Balances and Scales
The Explorer® series of semi-micro balances has been designed with the technology to ensure that very specific weighing results are accurate
  • Auto internal calibration
Powered by SmarText™ 2.0 software, Explorer® operates in the same manner as common electronics, with an icon-based touchscreen display, advanced applications, memory library, a USB host port, and more. A USB host port provides the capability of using a USB extension cable to directly transfer the data to a flash drive. The balance’s memory capabilities includes a library with space for up to 99 items, and a 10 recipe library for formulation mode. The built-in ionizer, included in Explorer automatic door models, generates bipolar ions continuously from positive and negative discharge electrodes and directs the ionized air onto the charged body to eliminate static electricity. These charges can build up in the weighing chamber and alter weighing results by as much as several milligrams.
Sophisticated laboratories requiring accuracy and looking for innovative technology to obtain measurement results can find both in Explorer® semi-micro balances. The series includes models with capacities up to 220g and resolutions up to 0.01mg, has been designed to use a High Speed Single Module weighing cell to ensure the accuracy of results. To minimize weighing errors and ensure accurate measurements, AutoCal™ utilizes two internal weights to perform a linear calibration.
Certifications: CE, FCC, CMC, OIML Class I.
Ordering information: All units are supplied with switching AC adapter with multiple plug head type for country, weighing pan, pan support, multi-language operator manual with quick start guide, and in-use cover. Terminal is included in box. All units are also supplied with a two-year manufacturer's warranty. Accessories not included. Draftshield is a standard accessory. Tower Mount is an optional accessory.