CYBRFast™ qPCR Lo-ROX Master Mix, Tonbo Biosciences
Supplier: Tonbo Biosciences
31-5101-0100R 31-5101-0500R
10002-224EA 118.5 CAD
10002-224 10002-226
CYBRFast™ qPCR Lo-ROX Master Mix, Tonbo Biosciences
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
CYBRFast qPCR Lo-ROX Master Mix is a ready-to-use cocktail for the amplification and detection of DNA on real-time instruments that support normalization with a standard ROX passive reference dye at a low final concentration of 50 nM
CYBRFast GREEN DNA intercalating dye does not inhibit PCR, resulting in better fluorescent signals than other dyes, and is optimized for use with CYBRFast Hot Start Taq DNA Polymerase. Novel hot start technology prevents primer dimer formation and non-specific amplification resulting in higher yields and specificity. Advanced buffer chemistry means little to no protocol optimization, regardless of template.
CYBRFast qPCR Master Mixes is used in real-time applications for improved signal to noise ratios, rapid extension rates for early Ct values, and enhanced sensitivity. This mix is suitable for high-throughput qPCR, and can be used with a variety of complex targets including genomic, cDNA and viral sequences - Please refer to our Instrument Compatibility Guide to ensure you have the correct ROX concentration for your system.
It reliably delivers enhanced fluorescence, speed, sensitivity and linearity.
Caution: Storage temperature: At -20°C upon Arrival and Limit Exposure to Light