VerityMAX™ DNA Polymerase, Tonbo Biosciences
Supplier: Tonbo Biosciences
31-5030-0250U 31-5030-1000U
10002-216EA 372.88 CAD
10002-216 10002-218
VerityMAX™ DNA Polymerase, Tonbo Biosciences
Nucleic Acid Reagents End-point PCR Enzymes and Kits
VerityMAX DNA Polymerase is a proprietary blend of our VerityPfu and Classic++ Hot Start Taq DNA polymerases, engineered and optimized specifically for generating amplicons of up to 35 kb in length with 2.5x higher fidelity than standard Taq polymerase
Resultant PCR products are both A-tailed and blunt-end finished. For added convenience and value, the advanced reaction buffer (5X) includes dNTPs and MgCl2. This enzyme is suitable for a wide variety of fast or standard PCR protocols when additional speed, sensitivity, specificity and yields are desired.
VerityMAX DNA Polymerase accurately and reliably amplifies DNA from complex GC/AT rich genomic DNA, plasmid templates and crude samples. It is suitable for multiplexing and colony PCR.
Hot start technology improves yields by minimizing or eliminating primer dimer formation and non-specific amplification