Classic++™ Hot Start Taq DNA Polymerase Master Mix, Tonbo Biosciences
Supplier: Tonbo Biosciences
31-5011-0200R 31-5011-1000R
10002-208EA 173.8 CAD
10002-208 10002-210
Classic++™ Hot Start Taq DNA Polymerase Master Mix, Tonbo Biosciences
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Classic++ Hot Start Taq DNA Polymerase Master Mix is a premixed, ready-to-use cocktail optimized for enhanced speed, yield and specificity.
This Master Mix contains an ultra-sensitive next generation recombinant Taq polymerase. Hot Start technology specifically blocks polymerase activity below 70°C and improves yields by minimizing or eliminating primer dimer formation and non-specific amplification.
Classic++ Hot Start Taq DNA Polymerase Master Mix is used for genotyping, colony screening, library construction and multiplex PCR applications under fast or standard cycling conditions. Classic++ Hot Start Polymerase allows for efficient amplification from complex templates and ultra-sensitive detection of low copy number templates.