Classic++™ Hot Start Taq DNA Polymerase, Tonbo Biosciences
Supplier: Tonbo Biosciences
31-5010-0250U 31-5010-1000U
10002-204EA 170.64 CAD
10002-204 10002-206
Classic++™ Hot Start Taq DNA Polymerase, Tonbo Biosciences
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Classic++ Hot Start Taq DNA Polymerase is an ultra-sensitive next generation recombinant Taq polymerase.
Hot Start technology specifically blocks polymerase activity below 70°C and improves yields by minimizing or eliminating primer dimer formation and non-specific amplification.
For added convenience and value, the advanced Reaction Buffer (5X) includes dNTPs and MgCl2. Classic++ Hot StartTaq DNA Polymerase is used for genotyping, colony screening, library construction and multiplex PCR applications under fast or standard cycling conditions. Hot Start polymerase allows for efficient amplification from complex templates and ultra-sensitive detection of low copy number templates.