Classic++™ Taq DNA Polymerase Master Mix, Tonbo Biosciences
Supplier: Tonbo Biosciences
31-5001-0200R 31-5001-1000R
10002-200EA 143.78 CAD
10002-200 10002-202
Classic++™ Taq DNA Polymerase Master Mix, Tonbo Biosciences
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Classic++ Taq DNA Polymerase Master Mix is a premixed, ready-to-use cocktail, optimized for efficient amplification of DNA templates by PCR.
Classic++ Taq DNA Polymerase is a robust next generation recombinant Taq polymerase optimized for exceptional everyday PCR. Convenient and reliable, Classic++ Taq polymerase is suitable for routine PCR protocols and has been engineered to provide enhanced speed, yield and specificity over that of standard Taq polymerase. Use Classic++ Taq DNA Polymerase Master Mix for high-throughput PCR, TA cloning, genotyping, library construction and RT-PCR under fast or standard cycling conditions.
Works well with crude samples and complex template DNA.