VWR® Screw Thread Vial with Cap
Supplier: VWR International
66010-152 66010-156
66010-152CS 2940.14 CAD
66010-152 66010-156
VWR® Screw Thread Vial with Cap
Vials
Made of type 1 clear borosilicate glass, these screw thread vials are designed with a continuous thread 'CT' finish, for a positive seal in a variety of uses including laboratory or manufacturer's use.
  • Borosilicate glass fabrication
  • Polypropylene cap
  • Screw thread vial design
Accompanied by a polypropylene foam lined polypropylene closed top cap.
Available in a wide range of sizes.
Ordering information: In attractive, white low-particulate cardboard boxes.