Corning® Rocker Cell Culture Bags, Corning

Supplier: Corning
91-200-78 91-200-79 91-200-80 91-200-81 91-200-92
CA89423-228EA 714.29 CAD
CA89423-228 CA89423-230 CA89423-232 CA89423-234 CA89508-740
Corning® Rocker Cell Culture Bags, Corning
Bags Media Bags
Rocker cell culture bags are designed for use with commercial rocking motion bioreactor chambers, and feature irradiated ethylene vinyl acetate (EVA)/low-density polyethylene (ULDPE) 9101 film and stability bars on each side. These sterile, single-use cell culture bags are ideal for applications from basic research to large-scale biopharmaceutical manufacturing.

  • Provides non-invasive mixing of the cell culture/media with efficient gas transfer
  • Eliminates the risk of cross-contamination
  • Used for basic research to large scale cell culture applications
  • Available in choice of sizes: 2, 10, 20, 22 and 50 L

The rocking motion of the bioreactor platform creates effective, non invasive mixing of the cell culture/media with efficient gas transfer. Rocking-motion prevents cells from settling at the bottom of the rocker cell culture bag while generating large amounts of recombinant protein in both mammalian and insect cell lines.

Port configurations:

Port 1 (all bag sizes)
Check valve with ¹/₄" barb
Pall 8004022 filter
2" length ³/₁₆" Int.Ø. × ³/₈" Ext.Ø. PCMGS tube

Port 2 (all bag sizes)
Pall 8004022 filter
2" length ³/₁₆" Int.Ø. × ³/₈" Ext.Ø. PCMGS tube

Port 3 (2 L, 10 L, 20 L, 50 L bags)
Female MPC connector with ¹/₄" barb
Male MPC plug
40" length ¹/₄" Int.Ø. × ⁷/₁₆" Ext.Ø. chemically resistant, heat sealable flexible tube

Port 3 (22 L bag only)
Female MPC connector with ¹/₄" barb
Male MPC plug
48" length ¹/₄" Int.Ø. × ⁷/₁₆" Ext.Ø. chemically resistant, heat sealable flexible tube

Port 4 (2 L, 10 L, 20 L, 50 L bags)
Female Luer with ¹/₄" barb
Male Luer plug
40" length ¹/₈" Int.Ø. × ¹/₄" Ext.Ø. chemically resistant, heat sealable flexible tube

Port 4 (22 L bag only)
Female Luer with ¹/₄" barb
Male Luer plug
48" length ¹/₈" Int.Ø. × ¹/₄" Ext.Ø. chemically resistant, heat sealable flexible tube

Port 5 (all bag sizes)
Female Luer with ¹/₄" barb
Swabbable valve
Male Luer plug without stem

Ordering information: Optional accessories and tubing sets are available.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR