Corning® Cell Expansion Bags, Corning
Supplier: Corning
91-200-84 91-200-85 91-200-86 91-200-87
CA89423-240EA 133.14 CAD
CA89423-240 CA89423-242 CA89423-244 CA89423-246
Corning® Cell Expansion Bags, Corning
Bags Media Bags
Corning® Cell Expansion Bags are intended for the expansion and culture of non-adherent cells.
  • Made from a single-web polyolefin, gas permeable film
The integrated tubing allows for functionally closed system filling, feeding, and sampling.